Anti-ESR1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128043-100 ORB128043-200
BIRBORB128043-100EA 0 EUR
BIRBORB128043-100 BIRBORB128043-200
Anti-ESR1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool ESR1
Type antilichaam Primair,Estrogen Receptor 1
Environmentally Preferable
Type antilichaam Primair,Estrogen Receptor 1
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
ImmunoChemie Yes
Modification phospho-Ser102
Purification purified by affinity chromatography
Concentratie 1 mg/ml
Opslagbuffer PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR