Anti-MED24 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128354-100 ORB128354-200
BIRBORB128354-100EA 0 EUR
BIRBORB128354-100 BIRBORB128354-200
Anti-MED24 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: MED24 (mediator complex subunit 24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR