Anti-ZDHHC2 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128930-100 ORB128930-200
BIRBORB128930-100EA 0 EUR
BIRBORB128930-100 BIRBORB128930-200
Anti-ZDHHC2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ZDHHC2 (zinc finger, DHHC-type containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR