Anti-Flag Tag Mouse monoclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB129610-100 ORB129610-20
BIRBORB129610-100EA 0 EUR
BIRBORB129610-100 BIRBORB129610-20
Anti-Flag Tag Mouse monoclonal antibody
Antilichamen
  • Western Blot: 1:3000-1:10000; Immunofluorescence: 1:200

Type: Primary
Antigen: Flag Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR