Anti-BCL10 Rabbit polyclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB136614-100 ORB136614-200 ORB136614-50
BIRBORB136614-100EA 0 EUR
BIRBORB136614-100 BIRBORB136614-200 BIRBORB136614-50
Anti-BCL10 Rabbit polyclonal antibody
Antilichamen
Type: Primary
Antigen: BCL10 (B-cell CLL/lymphoma 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR