Anti-DLGAP5 Rabbit polyclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB136668-100 ORB136668-200 ORB136668-50
BIRBORB136668-100EA 0 EUR
BIRBORB136668-100 BIRBORB136668-200 BIRBORB136668-50
Anti-DLGAP5 Rabbit polyclonal antibody
Antilichamen
Type: Primary
Antigen: DLGAP5 (discs, large (Drosophila) homolog-associated protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR