Anti-HSPA5 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB67506-200 ORB67506-50
BIRBORB67506-200EA 0 EUR
BIRBORB67506-200 BIRBORB67506-50
Anti-HSPA5 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:2000

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR