Anti-Hsp110 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORB67559-100 ORB67559-25
BIRBORB67559-100EA 0 EUR
BIRBORB67559-100 BIRBORB67559-25
Anti-Hsp110 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:1000
Type: Primary
Antigen: Hsp110
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now