Anti-CaMK2 alpha/ beta/ delta Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB99311-100 ORB99311-20
BIRBORB99311-100EA 0 EUR
BIRBORB99311-100 BIRBORB99311-20
Anti-CaMK2 alpha/ beta/ delta Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: CaMK2 alpha/ beta/ delta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR