Anti-IR Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB99411-100 ORB99411-20
BIRBORB99411-100EA 0 EUR
BIRBORB99411-100 BIRBORB99411-20
Anti-IR Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000

Type: Primary
Antigen: IR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR