Anti-ACTB Mouse Monoclonal Antibody [clone: mAbcam 8224]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB8224-100 AB8224-250
ABCAAB8224-100EA 475 EUR
ABCAAB8224-100 ABCAAB8224-250
Anti-ACTB Mouse Monoclonal Antibody [clone: mAbcam 8224]
Antilichamen
Opgelet:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone: mAbcam 8224
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now