Anti-LEF1 Rabbit Monoclonal Antibody [clone: EP2030Y]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB53293-100 AB53293-10
ABCAAB53293-100EA 725 EUR
ABCAAB53293-100 ABCAAB53293-10
Anti-LEF1 Rabbit Monoclonal Antibody [clone: EP2030Y]
Antilichamen
Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: LEF1 (lymphoid enhancer-binding factor 1)
Clonality: monoclonal
Clone: EP2030Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR