Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB76020-100 AB76020-250UL AB76020-20UL AB76020-1ML
ABCAAB76020-100EA 475 EUR
ABCAAB76020-100 ABCAAB76020-250UL ABCAAB76020-20UL ABCAAB76020-1ML
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y]
Antilichamen
Rabbit monoclonal [EP1845Y] to Sodium Potassium ATPase - Plasma Membrane Loading Control.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human;Chinese hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR