Anti-CAST Rabbit Monoclonal Antibody [clone: SP82]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB101684-100 AB101684-500 AB101684-1
ABCAAB101684-100EA 732 EUR
ABCAAB101684-100 ABCAAB101684-500 ABCAAB101684-1
Anti-CAST Rabbit Monoclonal Antibody [clone: SP82]
Antilichamen
Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: CAST (calpastatin)
Clonality: monoclonal
Clone: SP82
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR