Anti-Xanthine Oxidase Rabbit Monoclonal Antibody [clone: EPR4605]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB109235-100 AB109235-1ML
ABCAAB109235-100EA 682 EUR
ABCAAB109235-100 ABCAAB109235-1ML
Anti-Xanthine Oxidase Rabbit Monoclonal Antibody [clone: EPR4605]
Antilichamen
Rabbit monoclonal [EPR4605] to Xanthine Oxidase.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Xanthine Oxidase
Clonality: Monoclonal
Clone: EPR4605
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat