Anti-Xenotropic MuLV-related virus Rabbit Polyclonal Antibody

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB111014-100 AB111014-1
ABCAAB111014-100EA 500 EUR
ABCAAB111014-100 ABCAAB111014-1
Anti-Xenotropic MuLV-related virus Rabbit Polyclonal Antibody
Antilichamen
Rabbit polyclonal to Xenotropic MuLV-related virus.

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Xenotropic MuLV-related virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Xenotropic MuLV-related virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR