Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20037 20037-1
20037.EA 359 EUR
20037. 20037-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antilichamen
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse