Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20066 20066-1
20066.EA 315 EUR
20066. 20066-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Antilichamen
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR