Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20079-1 20079
20079-1EA 121 EUR
20079-1 20079.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Antilichamen
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken