Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20094 20094-1
20094.EA 359 EUR
20094. 20094-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)
Antilichamen
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR