Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20131 20131-1
20131.EA 238 EUR
20131. BTIU20131-1
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antilichamen
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF633
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR