Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20165 20165-1
20165.EA 300 EUR
20165. 20165-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Antilichamen
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken