Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20168 20168-1
20168.EA 359 EUR
20168. 20168-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Antilichamen
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR