Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20175-1 20175
20175-1EA 151 EUR
20175-1 20175.
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antilichamen
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse