Anti-IgG Donkey Polyclonal Antibody (Biotin)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20190 20190-1
20190.EA 359 EUR
20190. 20190-1
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antilichamen
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR