Anti-IgG Donkey Polyclonal Antibody (CF™ 680R)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20195 20195-1
20195.EA 368 EUR
20195. 20195-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 680R)
Antilichamen
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680R Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR