Anti-Biotin Mouse Monoclonal Antibody (CF®488A)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20204 20204-1
20204.EA 359 EUR
20204. 20204-1
Anti-Biotin Mouse Monoclonal Antibody (CF®488A)
Antilichamen
Type: Tag
Antigen: Biotin (biotin)
Clonality: monoclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR