Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20210 20210-1
20210.EA 385 EUR
20210. 20210-1
Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Tag
Antigen: Fluorescein (fluorescein)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR