Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20230 20230-1
20230.EA 359 EUR
20230. 20230-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antilichamen
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR