Anti-DAF-3 Rabbit Polyclonal Antibody
Leverancier: Novus Biologicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NB100-1924 NB100-1924SS
NOVUNB100-1924EA 554 EUR
NOVUNB100-1924 NOVUNB100-1924SS
Anti-DAF-3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: DAF-3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: C. elegans
Order Now
Specificaties
antigeen symbool DAF3
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity C. elegans
Host Rabbit
Gen-ID 180431
Isotype
Western blot Yes
ImmunoNeerslag Yes
ChIP Yes
Opslagtemperatuur Cool
Shipping temperature Ice