Anti-IRE1 alpha Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB100-2324 NB100-2324SS
NOVUNB100-2324EA 660 EUR
NOVUNB100-2324 NOVUNB100-2324SS
Anti-IRE1 alpha Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: IRE1 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR