Anti-CD36 Mouse Monoclonal Antibody [clone: D-2712]

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB110-59724 NB110-59724SS
NOVUNB110-59724EA 623 EUR
NOVUNB110-59724 NOVUNB110-59724SS
Anti-CD36 Mouse Monoclonal Antibody [clone: D-2712]
Antilichamen
The CD36 / SR-B3 Antibody (D-2712) from Novus Biologicals is a mouse monoclonal antibody to CD36 / SR-B3. This antibody reacts with mouse, rat, human (negative). The CD36 / SR-B3 Antibody (D-2712) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250

Type: Primary
Antigen: CD36/SR-B3
Clonality: Monoclonal
Clone: D-2712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat, Human (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR