Anti-NPM1 Rabbit Polyclonal Antibody
Leverancier: Novus Biologicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NB110-61646 NB110-61646SS
NOVUNB110-61646EA 638 EUR
NOVUNB110-61646 NOVUNB110-61646SS
Anti-NPM1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human