Anti-HIST3H3 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB21-1074 NB21-1074SS
NOVUNB21-1074EA 628 EUR
NOVUNB21-1074 NOVUNB21-1074SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR