Anti-CD36 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB400-144SS NB400-144
NOVUNB400-144SSEA 360 EUR
NOVUNB400-144SS NOVUNB400-144
Anti-CD36 Rabbit Polyclonal Antibody
Antilichamen
The CD36 / SR-B3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CD36 / SR-B3. This antibody reacts with human, mouse, rat, porcine, bovine, primate. The CD36 / SR-B3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200-1:400

Type: Primary
Antigen: CD36/SR-B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR