Anti-LRRK2 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NBP1-78299 NBP1-78299SS
NOVUNBP1-78299EA 554 EUR
NOVUNBP1-78299 NOVUNBP1-78299SS
Anti-LRRK2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR