Anti-SLC6A3 Mouse Monoclonal Antibody [clone: mAb16]
Leverancier: Novus Biologicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NBP2-22164 NBP2-22164SS
NOVUNBP2-22164EA 598 EUR
NOVUNBP2-22164 NOVUNBP2-22164SS
Anti-SLC6A3 Mouse Monoclonal Antibody [clone: mAb16]
Antilichamen
Type: Primary
Antigen: SLC6A3 (solute carrier family 6 (neurotransmitter transporter), member 3)
Clonality: monoclonal
Clone: mAb16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat