Anti-SLC2A2 Rabbit Polyclonal Antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NBP2-22218 NBP2-22218SS
NOVUNBP2-22218EA 642 EUR
NOVUNBP2-22218 NOVUNBP2-22218SS
Anti-SLC2A2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SLC2A2 (solute carrier family 2 (facilitated glucose transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR