Anti-Red Fluorescent Protein Rabbit Polyclonal Antibody
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1310-02 R1310-02-5
USBIR1310-02EA 377 EUR
USBIR1310-02 USBIR1310-02-5
Anti-Red Fluorescent Protein Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml
Type: Tag
Antigen: Red Fluorescent Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now