Anti-IgG Goat Polyclonal Antibody (Europium 1024)

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D17-114-50 D17-114-20 D17-114-1MG
COBSD17-114-50EA 1370 EUR
COBSD17-114-50 COBSD17-114-20 COBSD17-114-1MG
Anti-IgG Goat Polyclonal Antibody (Europium 1024)
Antilichamen
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR