Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (Europium 1024)
Leverancier: Columbia Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D17-1310-50 D17-1310-20 D17-1310-1MG
COBSD17-1310-50EA 1370 EUR
COBSD17-1310-50 COBSD17-1310-20 COBSD17-1310-1MG
Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (Europium 1024)
Antilichamen
Type: Tag
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now