Environmentally friendly, high contrast precision thermometers, BIO-TEMP®
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
515.060IMTJ-R 512A150IMTJ-R 5122P150-T-QP 5182T110-T-QP 5162T051-QP 5172T101-QP 5122T150-T-QP 5162T101-T-QP 5172T101-T-QP
620-0293EA 23.8 EUR
620-0293 ALLA512A150IMTJ-R 620-1495 620-0295 ALLA5162T051-QP ALLA5172T101-QP ALLA5122T150-T-QP ALLA5162T101-T-QP ALLA5172T101-T-QP
Environmentally friendly, high contrast precision thermometers, BIO-TEMP®
Thermometers Analoge thermometers
These precision thermometers are easy to read due to the high contrast blue or red graduations on a yellow background, and with an arrow printed at every 10 °C.
  • Accuracy without any of the hazards of mercury
  • Prtinted line for an immersion to the correct depth
  • Strong triangular ring moulded at the top to avoid rolling on a table
Filling: Blue or red liquid.
Leveringsinformatie: Supplied with certificate of traceability in a sturdy box.