Certified MS analysed vial kits
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5190-2278 5190-2280 5190-2279 5190-2281 5190-2282 5190-2283
HEWL5190-2278EA 112 EUR
HEWL5190-2278 HEWL5190-2280 HEWL5190-2279 HEWL5190-2281 HEWL5190-2282 HEWL5190-2283
Certified MS analysed vial kits
flesjes
Certified MS analysed vial kits end the need to pre-test or to rerun samples due to unexpected peaks. All MS analysed vials kits include a certificate of analysis that contains true, lot-specific and fully traceable LC/MS and GC/MS signal traces as well as critical physical dimensions.
  • Lot tested by both GC/MS and LC/MS
  • Larger write-on spots for easier labeling and identification
  • Packed in a clean environment using clean packaging to reduce contamination