Autosampler syringes for HP 7670/71/72 autosamplers
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5182-9622 5182-9626 5182-9734 5182-9799
HEWL5182-9622EA 105 EUR
HEWL5182-9622 HEWL5182-9626 HEWL5182-9734 HEWL5182-9799
Autosampler syringes for HP 7670/71/72 autosamplers
Spuiten Injectiespuiten voor chromatografie
HP7670/71/72 autosampler syringes feature a long needle and regular plunger button for compatibility with HP 7670/71/72 autosamplers.