Contact slides, BD Hycheck™
Leverancier: DIFCO BD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
290001 290002
DIFC290001EA 45.4 EUR
DIFC290001 DIFC290002
Contact slides, BD Hycheck™
Media voor microbiologie Geprepareerde media voor microbiologie
Hygiene contact slides used to assess the microbiological contamination of surfaces or fluids.
  • Double sided
  • Feature a hinged paddle that bends for easy sampling