High visibility trousers with laces, FARGO HV
Leverancier: DELTA PLUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FARGOHVJA1XL FARGOHVJA2XL FARGOHVJA3XL
DELPFARGOHVJA1XLEA 138 EUR
DELPFARGOHVJA1XL DELPFARGOHVJA2XL DELPFARGOHVJA3XL
High visibility trousers with laces, FARGO HV
Werkkleding
These trousers with waterproof seams are made of a polyurethane-coated Oxford polyester fabric. With wadded polyester Taffeta lining for protection from the cold and retro-reflective sewn bands for improved visibility.
  • Elasticated waist on both sides, adjustable elasticated braces
  • Preformed knees
  • Four pockets
  • Bottom of legs with gusset
  • Higher back protection
Certificaten: EN 14058 (332XX), EN 471 (1-2), EN 343 (3-1)