Hematocrit tubes, PLASTICRIT™
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8-000-7520/5 8-000-7520-H/5
DRUM8-000-7520/5EA 331 EUR
DRUM8-000-7520/5 DRUM8-000-7520-H/5
Hematocrit tubes, PLASTICRIT™
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These plastic hematocrit tubes provide the identical capillary action as glass hematocrit tubes. They eliminate the possibility of contamination due to breakage with no compromise in accuracy.
  • Available as plain or heparinised
Complies to FDA, NIOSH, CDC, and OSHA safety recommendations.