Trolleys for gas cylinders
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13265 13266
MSGA13265EA 354 EUR
MSGA13265 MSGA13266
Trolleys for gas cylinders
Transportwagens
These trolleys in PE can be used for gas cylinders of Ø upto 320 mm.
  • With tightening strap
  • With solid rubber tires (wheel Ø 200 mm)
  • Available with support wheel
  • 80 kg load capacity