Polyvinyl alcohol, M.W. 115,000 88, EMPROVE® EXPERT JPE, Ph. Eur., USP excipient_suitable, hydrolysed, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PVOH, PVA, Mowiol®
1.41353.1000 1.41353.9029
1.41353.1000EA 542 EUR
1.41353.1000 1.41353.9029
Polyvinyl alcohol, M.W. 115,000 88, EMPROVE® EXPERT JPE, Ph. Eur., USP excipient_suitable, hydrolysed, SAFC®
Polyvinyl alcohol
Formule: [-CH₂CH(OH)-]n
MDL Nummer: MFCD00081922
CAS nummer: 9002-89-5