Polyvinyl alcohol, M.W. 27,000 ≥98%, EMPROVE® EXPERT JPE suitable for use as excipient, hydrolysed, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.41356.1000 1.41356.9029
1.41356.1000EA 541 EUR
1.41356.1000 1.41356.9029
Polyvinyl alcohol, M.W. 27,000 ≥98%, EMPROVE® EXPERT JPE suitable for use as excipient, hydrolysed, SAFC®
Polyvinyl alcohol
Formule: [-CH₂CH(OH)-]n
MDL Nummer: MFCD00081922
CAS nummer: 9002-89-5